2-Chloropyrimidine ≥99.0%

Supplier: TCI America
Warning

C1050-25G
TCC1050-025GEA 87.09 USD
TCC1050-025G
2-Chloropyrimidine ≥99.0%
2-Chloropyrimidine

CAS Number: 1722-12-9
MDL Number: MFCD00006060
Molecular Formula: C4H3ClN2
Molecular Weight: 114.53
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 91
Melting point (°C): 63

Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 91 °C
Melting Pt: 63 °C
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/83/09/28938309.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR