α,α,α-Trifluoro-m-toluonitrile ≥98.0%

Supplier: TCI America

Synonyms: 3-(Trifluoromethyl)benzonitrile 3-Cyanobenzotrifluoride m-(Trifluoromethyl)benzonitrile

C1003-25G
TCC1003-025GEA 59.17 USD
TCC1003-025G
α,α,α-Trifluoro-m-toluonitrile ≥98.0%
α,α,α-Trifluoro-m-toluonitrile

CAS Number: 368-77-4
MDL Number: MFCD00001807
Molecular Formula: C8H4F3N
Molecular Weight: 171.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 72
Freezing point (°C): 17
Specific Gravity (20/20): 1.28


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Cyanobenzotrifluoride, alpha,alpha,alpha-Trifluoro-m-tolunitrile

Formula: C₈H₄F₃N
MW: 171.12 g/mol
Boiling Pt: 189 °C
Melting Pt: 19…22 °C
Density: 1.28 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00001807
CAS Number: 368-77-4
EINECS: 206-711-4
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/82/67/28938267.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR