β-Chloropropiophenone ≥96.0%
Supplier: TCI America
Synonyms:
3-Chloropropiophenone
C0995-25G
TCC0995-025GEA
84.28
USD
TCC0995-025G
β-Chloropropiophenone ≥96.0%
β-Chloropropiophenone
CAS Number: 936-59-4
MDL Number: MFCD00000990
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 48
Flash Point (°C): 113
Other Product Names / Synonyms: 3-Chloro-1-phenyl-1-propanone
Formula:
(C₆H₅)C(O)CH₂CH₂Cl MW: 168.62 g/mol Boiling Pt: 147 °C Melting Pt: 48 °C Density: 1.181 g/cm³ (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00000990 CAS Number: 936-59-4 EINECS: 213-317-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...