2-Chlorocyclohexanone ≥96.0% stabilized
Supplier: TCI America
Stabilization:
Stabilised with Stabilised
C0965-25G
TCC0965-025GEA
94.4
USD
TCC0965-025G
2-Chlorocyclohexanone ≥96.0% stabilized
2-Chlorocyclohexanone
(stabilized with HQ + CaCO3)
CAS Number: 822-87-7
MDL Number: MFCD00001626
Molecular Formula: C6H9ClO
Molecular Weight: 132.59
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 82
Freezing point (°C): 20
Specific Gravity (20/20): 1.16
Storage Temperature:
Formula:
C₆H₉ClO MW: 132.59 g/mol Boiling Pt: 107 °C Melting Pt: 18…20 °C Density: 1.16 (20 °C) Flash Pt: 82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001626 CAS Number: 822-87-7 EINECS: 212-505-5 UN: 3335 ADR: 9, |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...