Cyclopentadienylthallium ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Thallium(I) cyclopentadienide
C0954-5G
TCC0954-005GEA
205.03
USD
TCC0954-005G
Cyclopentadienylthallium ≥95.0% (by titrimetric analysis)
Cyclopentadienylthallium
CAS Number: 34822-90-7
MDL Number: MFCD00134141
Molecular Formula: C5H5Tl
Molecular Weight: 269.48
Purity/Analysis Method:
Form: Crystal
Formula:
C₅H₅Tl Boiling Pt: 90…110 °C (1 mmHg) Melting Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00134141 CAS Number: 34822-90-7 EINECS: 252-229-2 UN: 1707 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...