4-Chlorobutyronitrile ≥97.0%

Supplier: TCI America
Warning

C0925-500ML C0925-25ML
TCC0925-25MLEA 35.53 USD
TCC0925-25ML TCC0925-500ML
4-Chlorobutyronitrile ≥97.0%
4-Chlorobutyronitrile

CAS Number: 628-20-6
MDL Number: MFCD00001972
Molecular Formula: C4H6ClN
Molecular Weight: 103.55
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 108
Specific Gravity (20/20): 1.09

Formula: C₄H₆ClN
MW: 103.55 g/mol
Boiling Pt: 194 °C
Melting Pt: –44 °C
Density: 1.09 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00001972
CAS Number: 628-20-6
EINECS: 211-031-6
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/81/51/28938151.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR