2-Chloro-5-nitrobenzotrifluoride ≥97.0%

Supplier: TCI America

Synonyms: 1-Chloro-4-nitro-2-(trifluoromethyl)benzene 2-Chloro-5-nitrobenzotrifluoride

C0923-25G
TCC0923-025GEA 26.39 USD
TCC0923-025G
2-Chloro-5-nitrobenzotrifluoride ≥97.0%
2-Chloro-5-nitrobenzotrifluoride

CAS Number: 777-37-7
MDL Number: MFCD00007296
Molecular Formula: C7H3ClF3NO2
Molecular Weight: 225.55
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 232
Flash Point (°C): 98
Freezing point (°C): 22


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Chloro-4-nitro-2-(trifluoromethyl)benzene

Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 232 °C
Melting Pt: 25…27 °C
Density: 1.527 g/cm³ (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00007296
CAS Number: 777-37-7
EINECS: 212-287-1
https://us.vwr.com/stibo/web/std.lang.all/81/48/28938148.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR