Ethyl-4-chloroacetoacetate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl 4-chloro-3-oxobutanoate

C0911-500G C0911-25G
TCC0911-500GEA 157 USD
TCC0911-500G TCC0911-25G
Ethyl-4-chloroacetoacetate ≥95.0%
Ethyl 4-chloroacetoacetate

CAS Number: 638-07-3
MDL Number: MFCD00000939
Molecular Formula: C6H9ClO3
Molecular Weight: 164.59
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Melting point (°C): -8
Flash Point (°C): 96
Specific Gravity (20/20): 1.21


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: 4-Chloroacetoacetic Acid Ethyl Ester, Ethyl (Chloroacetyl)acetate, (Chloroacetyl)acetic Acid Ethyl Ester, Ethyl 4-Chloro-3-oxobutyrate, 4-Chloro-3-oxobutyric Acid Ethyl Ester

Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 120 °C
Melting Pt: –8 °C
Density: 1.21 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00000939
CAS Number: 638-07-3
EINECS: 211-317-0
https://us.vwr.com/stibo/web/std.lang.all/81/29/28938129.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR