Ethyl-4-chlorobutyrate ≥98.0%

Supplier: TCI America

C0830-25ML
TCC0830-025MLEA 17.26 USD
TCC0830-025ML
Ethyl-4-chlorobutyrate ≥98.0%
Ethyl 4-chlorobutyrate

CAS Number: 3153-36-4
MDL Number: MFCD00001004
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 80
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 4-Chlorobutyric Acid Ethyl Ester

Formula: C₆H₁₁ClO₂
Boiling Pt: 186 °C
Density: 1.08 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00001004
CAS Number: 3153-36-4
EINECS: 221-591-3
https://us.vwr.com/stibo/web/std.lang.all/79/10/28937910.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR