3-Picolyl chloride hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Chloromethyl)pyridine hydrochloride3-(Chloromethyl)-pyridinium chloride

C0825-100G C0825-25G
TCC0825-100GEA 255 USD
TCC0825-100G TCC0825-025G
3-Picolyl chloride hydrochloride ≥98.0% (by HPLC, titration analysis)
3-Picolyl chloride hydrochloride

CAS Number: 6959-48-4
MDL Number: MFCD00012818
Molecular Formula: C6H6ClN
Molecular Weight: 164.03
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144


Other Product Names / Synonyms: 3-Picolyl Chloride Hydrochloride

Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/78/74/28937874.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR