Methyl-4-chlorobutyrate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-chlorobutyrate

C0823-25ML
TCC0823-025MLEA 25.31 USD
TCC0823-025ML
Methyl-4-chlorobutyrate ≥98.0%
Methyl 4-chlorobutyrate

CAS Number: 3153-37-5
MDL Number: MFCD00001003
Molecular Formula: C5H9ClO2
Molecular Weight: 136.58
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 59
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 4-Chlorobutyric Acid Methyl Ester

Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 174 °C
Density: 1.13 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00001003
CAS Number: 3153-37-5
EINECS: 221-592-9
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/78/66/28937866.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR