Cyclooctanol ≥98.0%
Supplier: TCI America
|
Warning
|
C0623-25G
TCC0623-025GEA
123.83
USD
TCC0623-025G
Cyclooctanol ≥98.0%
Cyclooctanol
CAS Number: 696-71-9
MDL Number: MFCD00001744
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 88
Freezing point (°C): 17
Specific Gravity (20/20): 0.97
Formula:
C₈H₁₆O MW: 128.21 g/mol Boiling Pt: 108 °C Melting Pt: 15 °C Density: 0.97 (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001744 CAS Number: 696-71-9 EINECS: 211-800-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...