Carboxymethyl cellulose sodium salt n=∼1,050
Supplier: TCI America
Synonyms:
Carboxymethyl cellulose, sodium salt
, Cellulose, carboxymethyl ether, sodium salt
, CMC
, Cellulose gum
, Sodium carboxymethyl cellulose
C0603-500G
C0603-25G
TCC0603-025GEA
24.18
USD
TCC0603-025G
TCC0603-500G
Carboxymethyl cellulose sodium salt n=∼1,050
Carboxymethyl cellulose sodium salt
CAS Number: 9004-32-4
MDL Number: MFCD00081472
Form: Crystal
Color: White
Other Product Names / Synonyms: Cellulose Glycolic Acid Sodium Salt, Sodium Carboxymethyl Cellulose, Sodium Cellulose Glycolate, Sodium Tylose, Tylose Sodium
Formula:
[C₆H₇O₂(OH)x(OCH₂COONa)y]n Storage Temperature: Ambient |
MDL Number:
MFCD00081472 CAS Number: 9004-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...