2-Chlorobenzotrichloride ≥98.0%

Supplier: TCI America

Synonyms: 2-Chlorobenzotrichloride1-Chloro-2-(trichloromethyl)benzene

C0586-25G C0586-500G
TCC0586-25GEA 42 USD
TCC0586-25G TCC0586-500G
2-Chlorobenzotrichloride ≥98.0%
α,α,α,2-Tetrachlorotoluene

CAS Number: 2136-89-2
MDL Number: MFCD00000788
Molecular Formula: C7H4Cl4
Molecular Weight: 229.91
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 98
Freezing point (°C): 28


Other Product Names / Synonyms: 1-Chloro-2-(trichloromethyl)benzene, alpha,alpha,alpha,2-Tetrachlorotoluene

Formula: C₇H₄Cl₄
MW: 229.92 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 29…31 °C
Density: 1.5187 g/cm³ (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00000788
CAS Number: 2136-89-2
EINECS: 218-377-7
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/76/19/28937619.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR