Chlorocyclopentane ≥98.0%
Supplier: TCI America
Synonyms:
Cyclopentyl chloride
C0566-500ML
C0566-25ML
TCC0566-500MLEA
118.76
USD
TCC0566-500ML
TCC0566-25ML
Chlorocyclopentane ≥98.0%
Chlorocyclopentane
CAS Number: 930-28-9
MDL Number: MFCD00001360
Molecular Formula: C5H9Cl
Molecular Weight: 104.58
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 15
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: Cyclopentyl Chloride
Formula:
C₅H₉Cl MW: 104.58 g/mol Boiling Pt: 114 °C Melting Pt: –93 °C Density: 1.01 (20 °C) Flash Pt: 15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001360 CAS Number: 930-28-9 EINECS: 213-212-5 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...