Cyclopentene ≥98.0%

Supplier: TCI America

C0511-25ML C0511-500ML
TCC0511-25MLEA 23 USD
TCC0511-25ML TCC0511-500ML
Cyclopentene ≥98.0%
Cyclopentene

CAS Number: 142-29-0
MDL Number: MFCD00001394
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 46
Melting point (°C): -135
Flash Point (°C): -29
Specific Gravity (20/20): 0.77
Storage Temperature: 0-10°C

Formula: C₅H₈
Boiling Pt: 46 °C
Melting Pt: –135 °C
Density: 0.77 (20 °C)
Flash Pt: –29 °C
Storage Temperature: Ambient
MDL Number: MFCD00001394
CAS Number: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/75/39/28937539.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR