Cyclododecanol ≥98.0%

Supplier: TCI America

C0461-500G C0461-25G
TCC0461-025GEA 27.5 USD
TCC0461-025G TCC0461-500G
Cyclododecanol ≥98.0%
Cyclododecanol

CAS Number: 1724-39-6
MDL Number: MFCD00003717
Molecular Formula: C12H24O
Molecular Weight: 184.32
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 104
Melting point (°C): 77
Flash Point (°C): 138

Formula: C₁₂H₂₄O
MW: 184.32 g/mol
Boiling Pt: 104 °C
Melting Pt: 77 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00003717
CAS Number: 1724-39-6
EINECS: 217-031-2
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/74/83/28937483.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR