Cyanoacetamide ≥98.0%

Supplier: TCI America

Synonyms: 2-Cyanoacetamide

C0437-500G C0437-25G
TCC0437-500GEA 91.35 USD
TCC0437-500G TCC0437-025G
Cyanoacetamide ≥98.0%
Cyanoacetamide

CAS Number: 107-91-5
MDL Number: MFCD00008024
Molecular Formula: C3H4N2O
Molecular Weight: 84.08
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 120
Flash Point (°C): 215

Formula: C₃H₄N₂O
MW: 84.08 g/mol
Melting Pt: 120 °C
Flash Pt: 215 °C
MDL Number: MFCD00008024
CAS Number: 107-91-5
EINECS: 203-531-8
https://us.vwr.com/stibo/web/std.lang.all/74/60/28937460.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR