2-Hydroxy-m-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Cresotic acid2-Hydroxy-3-methylbenzoic acid3-Methylsalicylic acid

C0408-100G C0408-25G C0408-500G
TCC0408-500GEA 255 USD
TCC0408-500G TCC0408-100G TCC0408-025G
2-Hydroxy-m-toluic acid ≥98.0% (by GC, titration analysis)
2-Hydroxy-m-toluic acid

CAS Number: 83-40-9
MDL Number: MFCD00002448
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: o-Cresotic Acid, 2-Hydroxy-3-methylbenzoic Acid, 2-Hydroxy-m-toluic Acid

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00002448
CAS Number: 83-40-9
EINECS: 201-473-8
Merck Index: 13,02609
https://us.vwr.com/stibo/web/std.lang.all/74/34/28937434.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR