Creatine (N-amidinosarcosine) hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Amidinosarcosine hydrate2-(1-Methylguanidino)acetic acid hydrateCreatine hydrate

C0396-25G C0396-500G C0396-100G
TCC0396-500GEA 119.74 USD
TCC0396-500G TCC0396-025G TCC0396-100G
Creatine (N-amidinosarcosine) hydrate ≥98.0% (by HPLC, titration analysis)
Creatine (N-amidinosarcosine) hydrate

CAS Number: 6020-87-7
MDL Number: MFCD00004282
Molecular Formula: C4H9N3O
Molecular Weight: 131.14
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 295

Formula: C₄H₉N₃O₂·xH₂O
MW: 131.14 g/mol (anhy)
Melting Pt: 295 °C
MDL Number: MFCD00004282
CAS Number: 6020-87-7
EINECS: 200-306-6
https://us.vwr.com/stibo/web/std.lang.all/74/22/28937422.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR