Triethyl citrate ≥99.0%
Supplier: TCI America
Synonyms:
Citric acid triethyl ester
C0367-25G
C0367-500G
TCC0367-025GEA
27.14
USD
TCC0367-025G
TCC0367-500G
Triethyl citrate ≥99.0%
Triethyl citrate
CAS Number: 77-93-0
MDL Number: MFCD00009201
Molecular Formula: C12H20O7
Molecular Weight: 276.29
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 294
Melting point (°C): -45
Flash Point (°C): 176
Specific Gravity (20/20): 1.14
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Citric Acid Triethyl Ester
Formula:
C₁₂H₂₀O₇ MW: 276.29 g/mol Boiling Pt: 294 °C Melting Pt: –45 °C Density: 1.14 (20 °C) Flash Pt: 176 °C |
MDL Number:
MFCD00009201 CAS Number: 77-93-0 EINECS: 201-070-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...