Cinnamonitrile ≥95.0%

Supplier: TCI America

Synonyms: Cinnamonitrile3-Phenylacrylonitrile

C0361-25G
TCC0361-025GEA 61.27 USD
TCC0361-025G
Cinnamonitrile ≥95.0%
Cinnamonitrile

CAS Number: 1885-38-7
MDL Number: MFCD00001930
Molecular Formula: C9H7N
Molecular Weight: 129.16
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 113
Freezing point (°C): 19
Specific Gravity (20/20): 1.03


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Styryl Cyanide

Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 255 °C
Density: 1.03 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001930
CAS Number: 1885-38-7
EINECS: 224-441-5
https://us.vwr.com/stibo/web/std.lang.all/73/95/28937395.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR