2-Chloro-α,α,α-trifluorotoluene ≥98.0%

Supplier: TCI America

Synonyms: 1-Chloro-2-(trifluoromethyl)benzene 2-Chlorobenzotrifluoride

C0303-25G C0303-500G
TCC0303-500GEA 122.82 USD
TCC0303-500G TCC0303-25G
2-Chloro-α,α,α-trifluorotoluene ≥98.0%
2-Chloro-α,α,α-trifluorotoluene

CAS Number: 88-16-4
MDL Number: MFCD00000561
Molecular Formula: C7H4ClF3
Molecular Weight: 180.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 59
Specific Gravity (20/20): 1.37


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 1-Chloro-2-(trifluoromethyl)benzene, 2-Chloro-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 153 °C
Melting Pt: –5 °C
Density: 1.37 (20 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00000561
CAS Number: 88-16-4
EINECS: 201-805-1
UN: 2234
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/73/46/28937346.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR