3-Chlorotoluene ≥99.0%

Supplier: TCI America
Warning

C0296-25G C0296-500G
TCC0296-25GEA 23.45 USD
TCC0296-25G TCC0296-500G
3-Chlorotoluene ≥99.0%
3-Chlorotoluene

CAS Number: 108-41-8
MDL Number: MFCD00000595
Molecular Formula: C7H7Cl
Molecular Weight: 126.58
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Melting point (°C): -48
Flash Point (°C): 53
Specific Gravity (20/20): 1.07

Formula: C₇H₇Cl
MW: 126.59 g/mol
Boiling Pt: 162 °C
Melting Pt: –48 °C
Density: 1.07 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00000595
CAS Number: 108-41-8
EINECS: 203-580-5
UN: 2238
ADR: 3,III
Merck Index: 13,02190
https://us.vwr.com/stibo/web/std.lang.all/73/42/28937342.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR