4-Chloro-2-isopropyl-5-methylphenol ≥99.0%

Supplier: TCI America

Synonyms: 6-Chlorothymol

C0294-25G
TCC0294-025GEA 96.8 USD
TCC0294-025G
4-Chloro-2-isopropyl-5-methylphenol ≥99.0%
4-Chloro-2-isopropyl-5-methylphenol

CAS Number: 89-68-9
MDL Number: MFCD00002326
Molecular Formula: C10H13ClO
Molecular Weight: 184.66
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 62


Other Product Names / Synonyms: 6-Chlorothymol

Formula: C₁₀H₁₃ClO
MW: 184.67 g/mol
Boiling Pt: 143 °C
Melting Pt: 62 °C
Density: 1.228 g/cm³ (20 °C)
MDL Number: MFCD00002326
CAS Number: 89-68-9
EINECS: 201-930-1
Merck Index: 13,02189
https://us.vwr.com/stibo/web/std.lang.all/73/40/28937340.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR