(±)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (±)-2-Chloropropanoic acid2-Chloropropionic acid

C0275-25G C0275-500G
TCC0275-500GEA 127.22 USD
TCC0275-500G TCC0275-25G
(±)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)
(±)-2-Chloropropionic acid

CAS Number: 598-78-7
MDL Number: MFCD00004224
Molecular Formula: C3H5ClO2
Molecular Weight: 108.52
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 186
Melting point (°C): -12
Flash Point (°C): 106
Specific Gravity (20/20): 1.26

Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 186 °C
Melting Pt: –12 °C
Density: 1.26 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00004224
CAS Number: 598-78-7
EINECS: 209-952-3
UN: 2511
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/73/21/28937321.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR