1-Chloro-2-propanol ≥70.0% (contains ca. 25% 2-Chloro-1-propanol)

Supplier: TCI America

Synonyms: 1-Chloropropan-2-olChloropropanolPropylene chlorohydrin

C0269-25ML C0269-500ML
TCC0269-25MLEA 29.44 USD
TCC0269-25ML TCC0269-500ML
1-Chloro-2-propanol ≥70.0% (contains ca. 25% 2-Chloro-1-propanol)
1-Chloro-2-propanol

CAS Number: 127-00-4
MDL Number: MFCD00004530
Molecular Formula: C3H7ClO
Molecular Weight: 94.54
Purity/Analysis Method: 70.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 52
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Propylene Chlorohydrin

Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 127 °C
Density: 1.11 (20 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00004530
CAS Number: 127-00-4
EINECS: 204-819-6
UN: 2929
ADR: 6.1,II
Merck Index: 12,08036
https://us.vwr.com/stibo/web/std.lang.all/73/17/28937317.jpg

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR