4-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-(4-Chlorophenyl)hydrazine hydrochloride4-Chlorophenylhydrazine hydrochloride

C0257-25G C0257-250G
TCC0257-025GEA 96.43 USD
TCC0257-025G TCC0257-250G
4-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
4-Chlorophenylhydrazinium chloride

CAS Number: 1073-70-7
MDL Number: MFCD00012943
Molecular Formula: C6H7ClN2
Molecular Weight: 179.04
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9
https://us.vwr.com/stibo/web/std.lang.all/73/08/28937308.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR