(2-Chlorophenoxy)acetic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-CPA
C0249-25G
TCC0249-025GEA
159.93
USD
TCC0249-025G
(2-Chlorophenoxy)acetic acid ≥99.0% (by GC, titration analysis)
(2-Chlorophenoxy)acetic acid
CAS Number: 614-61-9
MDL Number: MFCD00004298
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 149
Formula:
(C₆H₄Cl)OCH₂COOH MW: 186.59 g/mol Boiling Pt: 308 °C (1013 hPa) Melting Pt: 149 °C Density: 1.509 g/cm³ (20 °C) |
MDL Number:
MFCD00004298 CAS Number: 614-61-9 EINECS: 210-388-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...