2-Chloro-6-nitrotoluene ≥99.0%

Supplier: TCI America

C0230-500G C0230-25G
TCC0230-25GEA 25 USD
TCC0230-25G TCC0230-500G
2-Chloro-6-nitrotoluene ≥99.0%
2-Chloro-6-nitrotoluene

CAS Number: 83-42-1
MDL Number: MFCD00007205
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 241
Flash Point (°C): 125
Freezing point (°C): 36

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 241 °C
Melting Pt: 35…37 °C
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00007205
CAS Number: 83-42-1
EINECS: 201-475-9
UN: 3457
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/72/84/28937284.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR