o-Cresolphthalein 0.04% in ethanol 95%

Danger

Synonyms: 3,3-Bis(4-hydroxy-3-methylphenyl)isobenzofuran-1(3H)-one

C0218-100ML
TCC0218-100MLEA 48.41 USD
TCC0218-100ML
o-Cresolphthalein 0.04% in ethanol 95%
o-Cresolphthalein

[for pH Determination]
CAS Number: 596-27-0
MDL Number: MFCD00005912
Molecular Weight: 346.38
Flash Point (°C): 12

Formula: C₂₂H₁₈O₄
MW: 346.39 g/mol
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00005912
CAS Number: 596-27-0
EINECS: 209-881-8
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR