4-Chloro-2-nitroaniline ≥98.0%

Supplier: TCI America
Danger

C0215-25G C0215-500G
TCC0215-500GEA 268.98 USD
TCC0215-500G TCC0215-25G
4-Chloro-2-nitroaniline ≥98.0%
4-Chloro-2-nitroaniline

CAS Number: 89-63-4
MDL Number: MFCD00007836
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 118
Flash Point (°C): 191

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 118 °C
Density: 1.37 g/cm³ (20 °C) (anhydr.)
Flash Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00007836
CAS Number: 89-63-4
EINECS: 201-925-4
UN: 2237
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/72/70/28937270.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR