1-Chlorohexane ≥95.0%

Supplier: TCI America

C0184-500ML C0184-25ML
TCC0184-500MLEA 180.55 USD
TCC0184-500ML TCC0184-25ML
1-Chlorohexane ≥95.0%
1-Chlorohexane

CAS Number: 544-10-5
MDL Number: MFCD00001018
Molecular Formula: C6H13Cl
Molecular Weight: 120.62
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 27
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Hexyl Chloride

Formula: C₆H₁₃Cl
MW: 120.62 g/mol
Boiling Pt: 135 °C
Melting Pt: –94 °C
Density: 0.872 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001018
CAS Number: 544-10-5
EINECS: 208-859-5
UN: 1993
ADR: 3,III
Merck Index: 13,02163
https://us.vwr.com/stibo/web/std.lang.all/72/47/28937247.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR