Chlormequat chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Chloroethyltrimethylammonium chloride Chlorocholine chloride (2-Chloroethyl)trimethylammonium chloride

C0172-25G C0172-500G
TCC0172-25GEA 60.9 USD
TCC0172-25G TCC0172-500G
Chlormequat chloride ≥98.0% (by titrimetric analysis)
Chlormequat chloride

CAS Number: 999-81-5
MDL Number: MFCD00011869
Molecular Formula: C5H13Cl2N
Molecular Weight: 158.07
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Bioscience, Other Plant Hormones
Other Product Names / Synonyms: (2-Chloroethyl)trimethylammonium Chloride

Formula: ClCH₂CH₂N(CH₃)₃Cl
MW: 158.07 g/mol
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00011869
CAS Number: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,02120
https://us.vwr.com/stibo/web/std.lang.all/72/38/28937238.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR