1-Chloro-2-phenylethane ≥99.0%

Supplier: TCI America

Synonyms: 2-Phenylethyl chloride(2-Chloroethyl)benzene2-(Chloroethyl)benzene

C0167-25G
TCC0167-025GEA 32.48 USD
TCC0167-025G
1-Chloro-2-phenylethane ≥99.0%
1-Chloro-2-phenylethane

CAS Number: 622-24-2
MDL Number: MFCD00000977
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 64
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Phenethyl Chloride, 2-Phenylethyl Chloride

Formula: ClCH₂CH₂C₆H₅
MW: 140.61 g/mol
Boiling Pt: 200 °C
Melting Pt: –60 °C
Density: 1.07 (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00000977
CAS Number: 622-24-2
EINECS: 210-725-6
https://us.vwr.com/stibo/web/std.lang.all/72/33/28937233.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR