4-Chloro-1-butanol ≥85.0% (contains varying amounts of Tetrahydrofuran)

Supplier: TCI America

Synonyms: 4-Chlorobutan-1-ol

C0144-25G C0144-500G
TCC0144-025GEA 69.22 USD
TCC0144-025G TCC0144-500G
4-Chloro-1-butanol ≥85.0% (contains varying amounts of Tetrahydrofuran)
4-Chloro-1-butanol

CAS Number: 928-51-8
MDL Number: MFCD00002967
Molecular Formula: C4H9ClO
Molecular Weight: 108.57
Purity/Analysis Method: 85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 52
Flash Point (°C): 36
Specific Gravity (20/20): 1.09
Storage Temperature: 0°C


Other Product Names / Synonyms: Tetramethylene Chlorohydrin

Formula: C₄H₉ClO
MW: 108.57 g/mol
Boiling Pt: 52 °C
Density: 1.09 (20 °C)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002967
CAS Number: 928-51-8
EINECS: 213-175-5
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/72/16/28937216.jpg

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR