2-Chlorobutane ≥99.0%

Supplier: TCI America

C0143-500ML C0143-25ML
TCC0143-25MLEA 37.54 USD
TCC0143-25ML TCC0143-500ML
2-Chlorobutane ≥99.0%
2-Chlorobutane

CAS Number: 78-86-4
MDL Number: MFCD00000871
Molecular Formula: C4H9Cl
Molecular Weight: 92.57
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Flash Point (°C): -10
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: sec-Butyl Chloride

Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68 °C
Melting Pt: –132 °C
Density: 0.87 (20 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000871
CAS Number: 78-86-4
EINECS: 201-151-7
UN: 1127
ADR: 3,II
Merck Index: 13,01560
https://us.vwr.com/stibo/web/std.lang.all/72/14/28937214.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR