1,1'-Carbonyldiimidazole ≥97.0% (by gravimetric analysis)

Danger

Synonyms: 1,1'-Carbonylbis-1H-imidazole 1,1'-Carbonylbis(imidazole) CDI N,N'-Carbonyldiimidazole

C0119-25G C0119-5G C0119-250G
TCC0119-5GEA 29.83 USD
TCC0119-5G TCC0119-250G TCC0119-25G
1,1'-Carbonyldiimidazole ≥97.0% (by gravimetric analysis)
1,1'-Carbonyldiimidazole

[Coupling Agent for Peptides Synthesis]
CAS Number: 530-62-1
MDL Number: MFCD00005286
Molecular Formula: C7H6N4O
Molecular Weight: 162.15
Purity/Analysis Method: 97.0% (W)
Form: Crystal
Melting point (°C): 118
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Phosgene & Thiophosgene Alternatives, Peptide Synthesis, Bioscience, Crosslinkers
Other Product Names / Synonyms: 1,1'-Carbonylbis-1H-imidazole, CDI

Formula: C₇H₆N₄O
MW: 162.15 g/mol
Melting Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00005286
CAS Number: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8,II
Merck Index: 13,01829

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR