2-Chloroanilinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Chloroaniline hydrochloride

C0114-25G
TCC0114-25GEA 35.53 USD
TCC0114-25G
2-Chloroanilinium chloride ≥98.0% (by HPLC, titration analysis)
2-Chloroanilinium chloride

CAS Number: 137-04-2
MDL Number: MFCD00053282
Molecular Formula: C6H6ClN
Molecular Weight: 164.03
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₆H₇Cl₂N
MDL Number: MFCD00053282
CAS Number: 137-04-2
EINECS: 205-274-7
UN: 2018
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/71/89/28937189.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR