Benzyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
N,N,N-Trimethyl-1-phenylmethanaminium bromide
B2980-25G
B2980-250G
TCB2980-25GEA
33.48
USD
TCB2980-25G
TCB2980-250G
Benzyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)
Benzyltrimethylammonium bromide
CAS Number: 5350-41-4
MDL Number: MFCD00011780
Molecular Formula: C10H16BrN
Molecular Weight: 230.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 229
Formula:
C₁₀H₁₆BrN MW: 230.15 g/mol Melting Pt: 229 °C |
MDL Number:
MFCD00011780 CAS Number: 5350-41-4 EINECS: 226-320-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...