Benzo[b]thiophene-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B2033-1G
TCB2033-001GEA
53.8
USD
TCB2033-001G
Benzo[b]thiophene-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Benzo[b]thiophene-2-carboxylic acid
CAS Number: 6314-28-9
MDL Number: MFCD00051636
Molecular Formula: C9H6O2S
Molecular Weight: 178.21
Purity/Analysis Method:
Form: Crystal
Formula:
C₉H₆O₂S MW: 178.22 g/mol |
MDL Number:
MFCD00051636 CAS Number: 6314-28-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...