TCIAB2001

B2001-1G B2001-5G B2001-25G
TCB2001-001GEA 54.54 USD
TCB2001-001G TCB2001-5G TCB2001-25G
TCIAB2001
CAS Number: 2113-57-7
MDL Number: MFCD00000082
Molecular Formula: C12H9Br
Molecular Weight: 233.11
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 301
Flash Point (°C): 110
Specific Gravity (20/20): 1.41

Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 3-Bromodiphenyl
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR