4-Bromobenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (4-Bromophenyl)(phenyl)methanonep-Bromobenzophenone

B1917-25G
TCB1917-025GEA 70.04 USD
TCB1917-025G
4-Bromobenzophenone ≥98.0%
4-Bromobenzophenone

CAS Number: 90-90-4
MDL Number: MFCD00000103
Molecular Formula: C13H9BrO
Molecular Weight: 261.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 176
Melting point (°C): 82

Formula: C₁₃H₉BrO
MW: 261.12 g/mol
Boiling Pt: 176 °C
Melting Pt: 82 °C
MDL Number: MFCD00000103
CAS Number: 90-90-4
EINECS: 202-024-9
https://us.vwr.com/stibo/web/std.lang.all/36/89/28933689.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR