5-Bromonicotinic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromo-3-pyridinecarboxylic acid3-Bromo-5-pyridine carboxylic acid5-Bromopyridine-3-carboxylic acid

B1818-25G
TCB1818-025GEA 59.89 USD
TCB1818-025G
5-Bromonicotinic acid ≥99.0% (by GC, titration analysis)
5-Bromonicotinic acid

CAS Number: 20826-04-4
MDL Number: MFCD00009783
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal
Melting point (°C): 185


Other Product Names / Synonyms: 5-Bromo-3-pyridinecarboxylic Acid

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00009783
CAS Number: 20826-04-4
EINECS: 244-065-5
https://us.vwr.com/stibo/web/std.lang.all/35/90/28933590.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR