4-tert-Butylbenzyl alcohol ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
(4-tert-Butylphenyl)methanol
, (4-(tert-Butyl)phenyl)methanol
B1761-5G
B1761-25G
TCB1761-5GEA
87.29
USD
TCB1761-5G
TCB1761-025G
4-tert-Butylbenzyl alcohol ≥97.0%
4-tert-Butylbenzyl alcohol
CAS Number: 877-65-6
MDL Number: MFCD00004662
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 238
Flash Point (°C): 87
Specific Gravity (20/20): 0.97
Formula:
C₁₁H₁₆O MW: 164.25 g/mol Boiling Pt: 238 °C Density: 0.97 (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004662 CAS Number: 877-65-6 EINECS: 212-894-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...