4-Bromo-m-xylene ≥75.0%

Supplier: TCI America
Warning

Synonyms: 2,4-Dimethylbromobenzene1-Bromo-2,4-dimethylbenzene

B1712-25G
TCB1712-025GEA 34 USD
TCB1712-025G
4-Bromo-m-xylene ≥75.0%
4-Bromo-m-xylene

CAS Number: 583-70-0
MDL Number: MFCD00000071
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: 75.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 79
Specific Gravity (20/20): 1.36


Other Product Names / Synonyms: 1-Bromo-2,4-dimethylbenzene

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 205 °C
Melting Pt: –1 °C
Density: 1.36 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00000071
CAS Number: 583-70-0
EINECS: 209-517-8
https://us.vwr.com/stibo/web/std.lang.all/34/64/28933464.jpg

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR