(±)-Methyl-2-bromobutyrate ≥97.0%

Supplier: TCI America

Synonyms: Methyl 2-bromobutyrate

B1462-25G B1462-500G
TCB1462-25GEA 25.38 USD
TCB1462-25G TCB1462-500G
(±)-Methyl-2-bromobutyrate ≥97.0%
(±)-Methyl 2-bromobutyrate

CAS Number: 3196-15-4
MDL Number: MFCD00009666
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 68
Specific Gravity (20/20): 1.41


Other Product Names / Synonyms: 2-Bromobutyric Acid Methyl Ester

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 172 °C
Density: 1.41 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00009666
CAS Number: 3196-15-4
EINECS: 221-699-0
https://us.vwr.com/stibo/web/std.lang.all/32/50/28933250.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR