(±)-Methyl-2-bromopropionate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-bromopropanoate
B1460-25G
B1460-500G
TCB1460-25GEA
26.41
USD
TCB1460-25G
TCB1460-500G
(±)-Methyl-2-bromopropionate ≥98.0%
(±)-Methyl 2-bromopropionate
CAS Number: 5445-17-0
MDL Number: MFCD00000143
Molecular Formula: C4H7BrO2
Molecular Weight: 167.00
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 51
Specific Gravity (20/20): 1.50
Product Categories: Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 2-Bromopropionic Acid Methyl Ester
Formula:
C₄H₇BrO₂ MW: 167 g/mol Boiling Pt: 144 °C Density: 1.50 (20 °C) Flash Pt: 51 °C |
MDL Number:
MFCD00000143 CAS Number: 5445-17-0 EINECS: 226-642-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...