α,α-Dichlorotoluene ≥95.0%

Synonyms: Benzal chloridealpha,alpha-DichlorotolueneBenzylidene chlorideDichlorophenylmethaneBenzylidene dichloride

B1414-500G B1414-25G B1414-100G
TCB1414-100GEA 85.23 USD
TCB1414-100G TCB1414-25G TCB1414-500G
α,α-Dichlorotoluene ≥95.0%
α,α-Dichlorotoluene

CAS Number: 98-87-3
MDL Number: MFCD00000829
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Melting point (°C): -16
Flash Point (°C): 93
Specific Gravity (20/20): 1.25


Other Product Names / Synonyms: Benzylidene Dichloride, alpha,alpha-Dichlorotoluene

Formula: (C₆H₅)CHCl₂
MW: 161.03 g/mol
Boiling Pt: 207 °C
Melting Pt: –16 °C
Density: 1.25 (20 °C)
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000829
CAS Number: 98-87-3
EINECS: 202-709-2
UN: 1886
ADR: 6.1,II
Merck Index: 13,01056
https://us.vwr.com/stibo/web/std.lang.all/32/13/28933213.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR