2-Bromobenzonitrile ≥98.0%
Supplier: TCI America
Synonyms:
o-Bromobenzonitrile
B1369-25G
B1369-10G
TCB1369-25GEA
107.59
USD
TCB1369-25G
TCB1369-010G
2-Bromobenzonitrile ≥98.0%
2-Bromobenzonitrile
CAS Number: 2042-37-7
MDL Number: MFCD00001772
Molecular Formula: C7H4BrN
Molecular Weight: 182.02
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 253
Melting point (°C): 54
Flash Point (°C): 110
Other Product Names / Synonyms: 1-Bromo-2-cyanobenzene
Formula:
C₇H₄BrN MW: 182.02 g/mol Boiling Pt: 253 °C Melting Pt: 54 °C Density: 1.496 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001772 CAS Number: 2042-37-7 EINECS: 218-045-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...